Clodinafop-propargyl ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: (R)-2-[4-(5-Chloro-3-fluoro-2-pyridinyloxy)phenoxy]propanoic acid 2-Propynyl ester, 2-Propynyl (R)-2-[4-(5-Chloro-3-fluoro-2-pyridinyloxy)phenoxy]propanoate

C3552-25G C3552-5G
TCC3552-25GEA 148 USD
TCC3552-25G TCC3552-5G
Clodinafop-propargyl ≥98.0% (by total nitrogen basis)
Clodinafop-propargyl
Formula: C₁₇H₁₃ClFNO₄
MDL Number: MFCD01632328
CAS Number: 105512-06-9
UN: 3077
ADR: 9,III
Merck Index: 14,02370

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 57.0 to 61.0 deg-C
Specific rotation [a]20/D +45.0 to +50.0 deg (C=2, Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR