Bentranil ≥98.0% (by GC)

Supplier: TCI America

B5739-1G B5739-250MG
TCB5739-1GEA 579 USD
TCB5739-1G TCB5739-250MG
Bentranil ≥98.0% (by GC)
Bentranil
Formula: C₁₄H₉NO₂
Boiling Pt: 192 °C
MDL Number: MFCD00043598
CAS Number: 1022-46-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR