(m-Acrylamidophenyl)boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-Acrylamidophenylboronic acid, (3-Acrylamidophenyl)boronic acid

A3199-1G A3199-5G
TCA3199-1GEA 129 USD
TCA3199-1G TCA3199-5G
(m-Acrylamidophenyl)boronic acid (contains varying amounts of Anhydride)
(m-Acrylamidophenyl)boronic acid
Formula: C₉H₁₀BNO₃
MW: 191 g/mol
MDL Number: MFCD09025755
CAS Number: 99349-68-5

Order Now

Specification Test Results

Appearance White to Amber powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR