4-(3-Nitrophenyl)-1,3-thiazol-2-amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Amino-4-(3-nitrophenyl)thiazole, 4-(3-Nitrophenyl)thiazol-2-amine

A3086-1G A3086-200MG
TCA3086-1GEA 144 USD
TCA3086-1G TCA3086-200MG
4-(3-Nitrophenyl)-1,3-thiazol-2-amine ≥98.0% (by GC, titration analysis)
4-(3-Nitrophenyl)-1,3-thiazol-2-amine
Formula: C₉H₇N₃O₂S
MW: 221.24 g/mol
MDL Number: MFCD00022455
CAS Number: 57493-24-0

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 187.0 to 191.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR