SB-366791 ≥98.0% (by GC)

Supplier: TCI America

B5762-100MG B5762-25MG
TCB5762-100MGEA 363 USD
TCB5762-100MG TCB5762-25MG
SB-366791 ≥98.0% (by GC)
SB-366791
Formula: C₁₆H₁₄ClNO₂
MDL Number: MFCD01033818
CAS Number: 472981-92-3

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR