tert-Butylphosphonic dichloride ≥98.0% (by GC)

Supplier: TCI America
Danger

B5729-1G B5729-5G
TCB5729-1GEA 51 USD
TCB5729-1G TCB5729-5G
tert-Butylphosphonic dichloride ≥98.0% (by GC)
tert-Butylphosphonic dichloride
Formula: C₄H₉Cl₂OP
Boiling Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012289
CAS Number: 4707-95-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Light yellow crystal to lump, wax
Purity (GC) min. 98.0 %
Melting point 120.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR