(3,3,3-Trifluoropropyl)dimethoxymethylsilane ≥98.0% (by GC)

Supplier: TCI America
Warning

D5328-25G D5328-5G
TCD5328-25GEA 95 USD
TCD5328-25G TCD5328-5G
(3,3,3-Trifluoropropyl)dimethoxymethylsilane ≥98.0% (by GC)
(3,3,3-Trifluoropropyl)dimethoxymethylsilane
Formula: C₆H₁₃F₃O₂Si
Boiling Pt: 85 °C
Flash Pt: 58 °C
MDL Number: MFCD00053175
CAS Number: 358-67-8
EINECS: 206-619-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR