5-Amino-1-methyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole

A3303-25G A3303-5G
TCA3303-25GEA 235 USD
TCA3303-25G TCA3303-5G
5-Amino-1-methyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
5-Amino-1-methyl-1H-pyrazole
Formula: C₄H₇N₃
Boiling Pt: 105 °C
MDL Number: MFCD00068156
CAS Number: 1192-21-8

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point gt. 69.0 and le 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR