Azetidine ≥98.0% (by GC)

Supplier: TCI America
Danger

A3257-5G A3257-1G
TCA3257-1GEA 140 USD
TCA3257-1G TCA3257-5G
Azetidine ≥98.0% (by GC)
Azetidine
Formula: C₃H₇N
Boiling Pt: 62 °C
Flash Pt: −21 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005165
EINECS: 207-963-8
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR