N,N-Dimethylformamide-[D₇]

Supplier: TCI America
Danger

Synonyms: N,N-Di[2H3]methyl[2H]formamideDMF-D7DMF-d7N,N-Dimethylformamide-[²H₇]Formic acid dimethylamide-D7Heptadeutero-N,N-dimethylformamideFormdimethylamide-D7N,N-Dimethylformamide-d7

D5233-10ML
TCD5233-10MLEA 994.5 USD
TCD5233-10ML
N,N-Dimethylformamide-[D₇]
N,N-Dimethylformamide-[D7]
Formula: DCOON(CD₃)₂
MW: 80.04 g/mol
Flash Pt: 58 °C
MDL Number: MFCD00003286
CAS Number: 4472-41-7
EINECS: 224-745-8
UN: 2265
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless clear liquid
Degree of deuteration min. 99.5 atom%D
Water max. 500 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR