Biochrom Ultrospec® 30 Cell Density Meter
Supplier: Harvard Bioscience
Biochrom Ultrospec® 30
80-3008-01
MSPP-80-3008EA 2306.15 USD
MSPP-80-3008
Biochrom Ultrospec® 30 Cell Density Meter
Density Meters
The Ultrospec 30 Cell density meter is compact and portable, with a unique standard curve functionality.
  • Compact and portable: Hand-held instrument with rechargeable battery for easy movement around the lab
  • Touchscreen display: Intuitive menu arrangement easily navigated using the colour trouchscreen
  • Instrument data output: Measurement data can be exported to a USB flash drive as common-separated value (.csv)
Creation of standard curves: Generate and save cell and condition specific standard curves using absorbance values or McFarland standards to use for future quantification.
Wavelength range: 600 nm