Anti-CD105 Mouse Monoclonal Antibody [clone: 43A3] (Alexa Fluor® 488)
Supplier: STEMCELL Technologies
60039AD.1 60039AD
MSPP-60039AD1EA 296.69 USD
MSPP-60039AD1 MSPP-60039AD
Anti-CD105 Mouse Monoclonal Antibody [clone: 43A3] (Alexa Fluor® 488)
Antibodies
Mouse monoclonal IgG1 antibody against human, mouse CD105 (endoglin), Alexa Fluor® 488-conjugated.
  • Hematopoietic stem
  • Progenitor cells
  • Mesenchymal stem
  • Progenitor cells
The 43A3 antibody reacts with CD105 (endoglin), an ~180 kDa cell surface glycoprotein which is a disulfide-bonded homodimer of ~90 kDa type I transmembrane subunits. CD105 is a component of the TGF-β receptor complex and is expressed by vascular endothelial smooth muscle cells, syncytiotrophoblasts of placenta and activated macrophages, and at relatively low levels by stromalMore Product Information