Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14] (APC)
Supplier: STEMCELL Technologies
60109AZ 60109AZ.1
MSPP-60109AZEA 515.92 USD
MSPP-60109AZ MSPP-60109AZ1
Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14] (APC)
Antibodies
Rat monoclonal IgG2a antibody against mouse CD62L (L-selectin), APC-conjugated.
  • T cells
The MEL-14 antibody reacts with murine CD62L (L-Selectin or LECAM-1), an ~74 - 95 kDa type I glycoprotein expressed on the surface of a majority of leukocytes. CD62L mediates leukocyte-endothelial cell interactions through its association with ligands such as CD34, glyCAM-1, MAdCAM-1 and PSGL-1. It functions to facilitate lymphocyte rolling on the vascular endothelium, trafficking of lymphocytes to the lymph nodes, and homing of lymphocytes and neutrophils to sitesMore Product Information