Sampling Bottles, Wide Mouth, HDPE, with Screw Caps
Supplier: Sampling Systems
8126H-30 8126H-500 8126H-125 8126H-60 8126H-250
77577-098EA 83.42 USD
77577-098 77577-104 77577-106 77577-100 77577-102
Sampling Bottles, Wide Mouth, HDPE, with Screw Caps
Bottles Plastic Bottles
These wide mouth bottles have a screw on cap and are ideal for transporting samples, laboratory work and general use in production. The wide opening means that these bottles are suitable for both powders and liquids.
  • HDPE body with PP lid
  • Robust, water tight construction
  • Translucent HDPE walls enable users to see the material inside
  • Wide mouth means they can be filled easily
  • Made from food grade and FDA compliant material
  • BSE/TSE free
Certifications: Conforms to the latest FDA and EU requirements. USP class VI compliant.