Stainless Steel Buckets, Graduated
Supplier: SAMPLING SYSTEMS LTD
A264-12L A264-15 A264-15L A264-6 A264-10 A264-10L A264-12
77577-786EA 45.74 USD
77577-786 77577-788 77577-790 77577-792 77577-780 77577-782 77577-784
Stainless Steel Buckets, Graduated
Buckets
Stainless steel, graduated buckets are ideal for use in food, chemical, and general industry.
  • 304 stainless steel construction
  • Skirtband gives extra protection to the base
  • Robust design
  • Easy to use
Graduated stainless steel buckets are made from heavy gauge stainless steel and are very durable. They are ideal for general-purpose factory work. The swing handle allows them to be lifted easily. Optional lids are available separately.
The 10, 12 and 15 litre buckets feature a skirt band around the base. This provides extra protection to the base and increases stability.