CLR Series™ Racked Pipette Tips for Universal Fit Pipettes
Supplier: Polycarbin
CLR Series™ Polycarbin®
1PT0020FOU 1PT1250FOU 1PT0010FOU 1PT0200FOU
77575-324CS 393.75 USD
77575-324 77575-326 77575-328 77575-330
CLR Series™ Racked Pipette Tips for Universal Fit Pipettes
Pipette Tips
Polycarbin's pipette tip rack and lid are made of 100% circular economy, post-consumer recycled content, combining high performance with a significantly reduced carbon footprint. Pipette tips do not contain recycled content, and are universal fit.
  • Rack and lid made from 100% circular economy, recycled content
  • Manufactured in an ISO 9001 compliant facility
  • Carries a smaller carbon footprint
  • DNase, RNase and Pyrogen-free
  • Free of Per- and polyfluoroalkyl substances (PFAS)
  • Sterile
Polycarbin's line of low-carbon lab products are designed for the sustainability-minded scientist. When designing this line of pipette tips, the focus is on the tip box or rack which is the most dense, carbon-intensive piece of plastic.
All tip boxes of any brand can be remanufactured and transformed into the next line of lab products using Polycarbin's alpha carbin, a mailback service.