RNase 4
Supplier: New England Biolabs (NEB)
M1284L M1284S
77532-254EA 392 USD
77532-254 77532-252
RNase 4
Enzymes
RNase 4 is a single-stranded RNA endonuclease that cleaves 3´ of uridine in uridine-purine sequences (cut sites: U/A and U/G). RNase 4 allows more targeted digestion of substrate RNA compared to single-nucleotide specific RNases like T1 (cut site: after G), RNase U2 (cut sites: after A and G), or bovine pancreatic RNase A (cut sites: after C and U). RNase 4 endonuclease activity tolerates uridine base modifications such as pseudo-, N1-methyl-pseudo-, dihydro-, and 5-methoxy-uridine species (Ψ, m1Ψ, D, and mo5U).
  • Endoribonuclease activity tolerates common RNA chemical modifications
  • Enhance analysis of mRNA 5′ cap identity and distribution
RNase 4 is a single-stranded endoribonuclease that cleaves RNA at uridine-purine (U/R) dinucleotide sites. Generate a larger population of uniquely mappable oligonucleotides for improved RNA sequence characterization by LC-MS/MS. Due to the chemical mechanism of RNase 4 endonucleolytic cleavage, product oligonucleotides contain heterogenous 3´ ends, where most species contain a linear 3´-phosphate or cyclic 2´,3´-phosphate.