Cryoprecipitate Bath, Model 9406, PolyScience
Supplier: Polyscience
6260MXCRY10C 6250MYCRY30E
13272-292EA 12848.71 USD
13272-292 13272-290
Cryoprecipitate Bath, Model 9406, PolyScience
Baths Cryoprecipitate Baths
Designed for the safe, reliable thawing of fresh frozen plasma and whole blood
  • Removable, center-hinged reservoir cover
  • 4 °C preset thawing temperature
  • Bright temperature and elapsed time displays
  • Calibration capability
  • Removable, high-efficiency fluid filter
  • Reservoir drain
  • Over-temperature and flow alarms
Bath includes a built-in timer for elapsed thawing time and a convenient calibration offset function for matching displayed temperature reading to a traceable standard. Built-in self-diagnostics instantly alert users to high temperatures, low liquid flow, and other conditions that may compromise thawing performance or AHF recovery. The reservoir drain valve, reusable high-efficiency liquid filter, and reusable air filter all help minimize maintenance downtime.
Bath provides proper conditions for the recovery of cryoprecipitated antihemophilic factor (AHF). Features a 52.2 L (13.8 gal.) reservoir capable of holding up to 24 units of frozen plasma or whole blood. The reservoir has a large, 33x52.4 cm (13x205/8") opening for easy loading and unloading and comes with a hinged, fully removable cover.
Certifications: cTUVus compliant.