Uristix® Urinalysis Reagent Test Strips, Siemens Healthineers
Supplier: Siemens Healthineers
10312569 10339520
10789-330CS 559.11 USD
10789-330 10789-332
Uristix® Urinalysis Reagent Test Strips, Siemens Healthineers
Clinical Diagnostic Supplies Clinical Reagents and Kits
Uristix® urinalysis test strips are ready to use upon removal from the bottle and may be read visually or read instrumentally using the CLINITEK Status®+ Urine Analyser.
  • Ready-to-use
  • May be read visually or using instrument
  • For use with CLINITEK Status®+
The strips are for professional, in vitro diagnostic use only.
Packaging: 100 strips per bottle.