OSOM® BVBlue® Test Kit, Sekisui Diagnostics
Supplier: SEKISUI DIAGNOSTIC
183 184
71001-056CS 2750.61 USD
71001-056 71001-058
OSOM® BVBlue® Test Kit, Sekisui Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Rapid test for the diagnosis of bacterial vaginosis. Detects elevated vaginal fluid sialidase activity, an enzyme produced by bacterial pathogens associated with bacterial vaginosis including Gardnerella, Bacteroides, Prevotella and Mobiluncus.
  • 92.8% sensitive, 98% specific versus Gram Stain
  • 1 minute hands-on-time; 10 minute read time
  • Instant color change provides clear easy-to-read results.
  • CPT Code: 87905QW
Detects all four major pathogens which cause BV. Test uses a vaginal swab; a speculum may not be required. Refrigerated storage at 2–8°C (35.6–46.4°F).
Certifications: CLIA-waived.
Ordering information: Control kit (sold separately) includes one positive and one negative control.