Genevac MV QHP Centrifugal Concentrator for Parallel Sample Evaporation
Supplier: ATS Scientific Products
Genevac
100409198
77574-998EA 20237.86 USD
77574-998
Genevac MV QHP Centrifugal Concentrator for Parallel Sample Evaporation
Concentrators
MV QHP-12060 Quattro high package.
  • Compact package, includes Duo concentrator, SpeedTrap condenser, Duo pump and connection kit, makes efficient use of bench space
  • Concentrator can accommodate medium sample capacities
  • Simple user interface with large digital display
  • Quiet electro-magnetic drive ensures maintenance free operation
  • Precise temperature control
  • Variety of solid aluminium sample rotors are available for tubes, micro tubes, vials or microplates and should be ordered separately
miVac centrifugal evaporators provide a compact, high performing and flexible solution for removing water and organic solvents from a variety of sample formats including tubes, micro tubes, microplates and vials. Ideal for life science laboratories, these centrifugal concentrators rely on the vacuum in the system to cause liquids to boil at a lower than normal temperature enabling concentrating or removal of solvents at very low temperatures.
To prevent evaporating samples from boiling over (bumping) in an uncontrolled manner, simultaneous to the drying process, samples are spun in a centrifuge creating sufficient g-force to keep each sample in its own tube.
The Quattro high package is designed for concentrating higher numbers of samples at one time. The Quattro concentrator has a larger capacity concentrator and also included are the Quattro vacuum pump, a quiet and compact four-head diaphragm pump, which can reach 3 mbar or below. Required for successful drying of medium boiling point solvents. Pump is housed in a case to match the concentrator.
The miVac SpeedTrap chills to −50 °C collecting solvents as a liquid in an easy to empty jar. The design and built-in compressor enable a small footprint while delivering big results.
Ordering information: Sample rotor must be ordered separately. A range of solid Quattro aluminium rotors are available to accommodate all common tubes, vials centrifuge tubes and micro centrifuge tubes. For the Quattro range a four position swing rotor is available.
Delivery information: Package includes Duo concentrator, SpeedTrap condenser, Duo pump and connection kit.
Caution: To keep your centrifugal evaporation system in good working condition, order consumable parts.