Genevac MV Freeze Drying Accessory Kit for MV Centrifugal Concentrator
Supplier: ATS Scientific Products
Genevac
100408209
77574-992EA 1493.33 USD
77574-992
Genevac MV Freeze Drying Accessory Kit for MV Centrifugal Concentrator
Concentrators
Use a miVac SpeedTrap as a stand-alone freeze dryer with miVac lyophilization accessories.
  • Can be used with stand-alone freeze drying accessory kit
  • Freeze up to 250 ml of water
The MV SpeedTrap can be used in conjunction with a Super Vacuum pump to freeze dry up to 250 ml of water. Pre-frozen samples can be placed in the cascade condenser jar provided with the freeze drying accessory kit.
A pressure controller is recommended when samples will be freeze dried using the concentrator to ensure optimal vacuum level for your process.
Ordering information: Freeze drying accessory kit includes, cascade condenser jar, a set of two holders, handle connection tubes and taps. Flasks must be ordered separately. Freeze drying accessories are not included and must be ordered separately.