Genevac MV SVP Super Vacuum Pump for MV Centrifugal Concentrator
Supplier: ATS Scientific Products
Genevac
100408450
77574-994EA 10049.06 USD
77574-994
Genevac MV SVP Super Vacuum Pump for MV Centrifugal Concentrator
Concentrators
miVac centrifugal evaporation systems provide a compact, high performing solution for removing water and organic solvents from a variety of sample formats. Configure a system to meet your application requirements.
  • Concentrators control the pump, heat and run time with a simple user interface and large digital display
  • Quiet electro-magnetic drive ensures maintenance free operation
  • Choose from two centrifuge bowl sizes, three pumps and add-on accessories
  • Variety of sample holder rotors, ordered separately accommodate tubes, micro tubes, vials or microplates
The Super Vacuum Pump is a scroll type vacuum pump for exceptionally demanding applications and freeze drying. This pump is capable of removing higher-boiling solvents at pressures down to 0.15 mbar.