VWR® Ultra-Micro Pipette Tips for Matrix Impact2™ Pipettors, Graduated
Supplier: VWR International
82018-040 82018-042
82018-040CS 1069.02 USD
82018-040 82018-042
VWR® Ultra-Micro Pipette Tips for Matrix Impact2™ Pipettors, Graduated
Pipette Tips
Sterile tips are graduated at 2 and 10 µl.
  • RNase- and DNase-free
  • Endotoxin-free
  • Fully autoclavable
  • Sterile
Tips and hinged boxes are fully autoclavable to 122 °C (252 °F).
Ordering information: Pak Rak® Express refills contain pipette tips to fill 8×12 racks, without the packaging waste of standard tip boxes. Pak Rak® Express refills require the Pak Rak® system loader (see 30128-374 series), which dispenses tips quickly and easily from the refill pack into each rack, eliminating risk of contamination from hands or gloves.