Owl™ Immobilon™-P Transfer Membranes, PVDF, Thermo Scientific
Supplier: Thermo Scientific
Owl™ Immobilon®
TM151-1 TM151-5 TM151-4
27373-528PK 547.84 USD
27373-528 27373-531 27373-532
Owl™ Immobilon™-P Transfer Membranes, PVDF, Thermo Scientific
Transfer Membranes
Hydrophobic polyvinylidene fluoride transfer membrane has a high signal-to-noise ratio, which allows for the use of mild blocking procedures
Membrane has high protein retention, is compatible with all standard visualization methods, and can be used for protein sequencing. Available in sheet or roll form.
Thermo Scientific offers Immobilon™-P, PVDF transfer membrane manufactured by Millipore™