VOA Vials - Solid Top Closures, Precleaned
Supplier: J.G. FINNERAN ASSOCIATES, INC.
9-090-2 9-120-2 9-121-2
77547-566CS 79.85 USD
77547-566 77547-568 77547-570
VOA Vials - Solid Top Closures, Precleaned
Vials
Precleaned VOA vials assembled with solid top white polypropylene closures, PTFE lined.
  • Available in clear or amber borosilicate glass
  • Assembled with solid top PTFE-lined closures
  • Non graduated
  • Non silanized
CLASS 2: (Precleaned) Containers are processed according to EPA recommended wash procedures. Washed with laboratory grade biodegradable, non-phosphate detergent, rinsed with ASTM Type I de-ionized water, oven dried, and assembled in a contaminate-free environment. The exterior of each case is labeled with a lot number.