Halogen Bulb, 12 V / 30 W, Leica Microsystems
Supplier: Leica Microsystems
11500317
10037-722EA 51.76 USD
10037-722
Halogen Bulb, 12 V / 30 W, Leica Microsystems
Lights Halogen Lights, Lamps and Bulbs
Replacement bulb for the following Leica microscopes: DM1000, DM2000, DM3000 and DMLS.