2-Bromo-3-fluoro-6-nitroaniline ≥98%

Supplier: Ambeed
Danger

A998423-1G A998423-250MG A998423-5G
77873-407EA 14 USD
77873-407 77873-408 77873-409
2-Bromo-3-fluoro-6-nitroaniline ≥98%
2-Bromo-3-fluoro-6-nitroaniline

Caution: for Research Use Only

Formula: C₆H₄BrFN₂O₂
MDL Number: MFCD18071077
CAS Number: 1218764-80-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR