Furan-2,5-dicarbonyl dichloride ≥98% (by GC)

Supplier: Ambeed
Danger

Synonyms: 2,5-Furandicarbonyl dichloride

A986379-25G A986379-5G
77573-950EA 89 USD
77573-950 77618-700
Furan-2,5-dicarbonyl dichloride ≥98% (by GC)
Furan-2,5-dicarbonyl dichloride

Caution: for Research Use Only

Formula: C₆H₂Cl₂O₃
MDL Number: MFCD28044847
CAS Number: 10375-34-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR