Ampicillin trihydrate ≥96%
Supplier: Ambeed
|
Danger
|
Synonyms:
(2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
A980131-100G
A980131-5G
A980131-500G
A980131-25G
77623-234EA
52.53
USD
77623-234
77623-236
77623-238
77784-472
77784-471
Ampicillin trihydrate ≥96%
Ampicillin trihydrate
Caution: for Research Use Only
Formula:
C₁₆H₁₉N₃O₄S·3H₂O MW: 403.45 g/mol |
MDL Number:
MFCD00072036 CAS Number: 7177-48-2 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...