R-(+)-IAA-94 ≥98%

Supplier: Ambeed
Warning

A886198-100MG A886198-10MG A886198-5MG A886198-250MG A886198-50MG
77670-286EA 530.28 USD
77670-286 77670-288 77670-290 77873-337 77873-338
R-(+)-IAA-94 ≥98%
R-(+)-IAA-94

Caution: for Research Use Only

Formula: C₁₇H₁₈Cl₂O₄
MDL Number: MFCD28133534
CAS Number: 54197-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR