Myrcene 75% stabilized

Supplier: Ambeed
Danger

Synonyms: 7-Methyl-3-methyleneocta-1,6-diene

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

A881518-25G
77670-280EA 29 USD
77670-280
Myrcene 75% stabilized
Myrcene

Caution: for Research Use Only

Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00008908
CAS Number: 123-35-3
UN: 2319
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR