(+)-Valencene 70%

Supplier: Ambeed
Danger

A839231-1G A839231-25G A839231-5G
77618-344EA 28.17 USD
77618-344 77618-346 77618-348
(+)-Valencene 70%
(+)-Valencene

Caution: for Research Use Only

Formula: C₁₅H₂₄
MDL Number: MFCD00075884
CAS Number: 4630-07-3
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR