Bacitracin ≥65 IU/mg

Supplier: Ambeed
Warning

A839108-10MG A839108-50MG A839108-1G A839108-250MG A839108-25G A839108-5G A839108-100G A839108-100MG
77595-336EA 5.13 USD
77595-336 77595-338 77603-130 77603-132 77603-134 77627-642 77878-687 77878-688
Bacitracin ≥65 IU/mg
Bacitracin

Caution: for Research Use Only

Formula: C₆₆H₁₀₃N₁₇O₁₆S₁
MW: 1421.60 g/mol
MDL Number: MFCD00062640
CAS Number: 1405-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR