1,2,3,4-Tetramethylcyclopentadiene ≥98% (mixture of isomers)

Supplier: Ambeed
Danger

Synonyms: 1,2,3,4-Tetramethyl-1,3-cyclopentadiene1,2,3,4-Tetramethylcyclopenta-1,3-dieneTetramethylcyclopentadiene

A834788-1G
77618-332EA 89.61 USD
77618-332
1,2,3,4-Tetramethylcyclopentadiene ≥98% (mixture of isomers)
1,2,3,4-Tetramethylcyclopentadiene

Caution: for Research Use Only

Formula: C₉H₁₄
MW: 122.21 g/mol
MDL Number: MFCD00145385
CAS Number: 4249-10-9
UN: 3295
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR