Cis-cyclooctane-1,2-diol ≥97%

Supplier: Ambeed
Danger

A819262-1G A819262-5G
77880-617EA 42 USD
77880-617 77880-618
Cis-cyclooctane-1,2-diol ≥97%
Cis-cyclooctane-1,2-diol

Caution: for Research Use Only

Formula: C₈H₁₆O₂
MDL Number: MFCD00134226
CAS Number: 27607-33-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR