Bromobimane (mBBr) ≥98%

Supplier: Ambeed
Danger

A805898-100MG A805898-50MG
77880-603EA 360 USD
77880-603 77880-604
Bromobimane (mBBr) ≥98%
Bromobimane (mBBr)

Caution: for Research Use Only

Formula: C₁₀H₁₁BrN₂O₂
MDL Number: MFCD00036951
CAS Number: 71418-44-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR