Titanyl(IV) acetylacetonate ≥98%
Supplier: Ambeed
|
Warning
|
A771193-25G
A771193-500G
A771193-100G
A771193-5G
77602-974EA
18.45
USD
77602-974
77670-064
77873-224
77873-225
Titanyl(IV) acetylacetonate ≥98%
Titanyl (IV) acetylacetonate
Caution: for Research Use Only
Formula:
C₁₀H₁₄O₅Ti |
MDL Number:
MFCD00013505 CAS Number: 14024-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...