(E)-8-(3-(4-Chlorophenyl)acryloyl)-3-(3,4-dichlorobenzyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one ≥98%

Supplier: Ambeed
Warning

A723809-10MG A723809-1MG A723809-5MG
77598-796EA 125.05 USD
77598-796 77598-798 77598-800
(E)-8-(3-(4-Chlorophenyl)acryloyl)-3-(3,4-dichlorobenzyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one ≥98%
(E)-8-(3-(4-Chlorophenyl)acryloyl)-3-(3,4-dichlorobenzyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one

Caution: for Research Use Only

Formula: C₂₃H₂₁Cl₃N₂O₃
MDL Number: MFCD30738004
CAS Number: 1334294-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR