5,5-Difluoro-1,3,7,9-tetramethyl-10-phenyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide ≥97%

Supplier: Ambeed
Warning

A720827-100MG A720827-250MG A720827-1G
77598-776EA 45.1 USD
77598-776 77622-560 77783-750
5,5-Difluoro-1,3,7,9-tetramethyl-10-phenyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide ≥97%
5,5-Difluoro-1,3,7,9-tetramethyl-10-phenyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide

Caution: for Research Use Only

Formula: C₁₉H₁₉BF₂N₂
MDL Number: MFCD28137455
CAS Number: 194235-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR