tert-Butyl (10-bromodecyl)carbamate ≥95%
Supplier: Ambeed
|
Danger
|
A693940-250MG
A693940-100MG
77573-246EA
164
USD
77573-246
77622-470
tert-Butyl (10-bromodecyl)carbamate ≥95%
tert-Butyl (10-bromodecyl)carbamate
Caution: for Research Use Only
Formula:
C₁₅H₃₀BrNO₂ |
MDL Number:
MFCD03788821 CAS Number: 887353-29-9 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...