Polyethylene glycol monododecyl ether Hydroxyl Value 40-60 mg KOH/g
Supplier: Ambeed
|
Danger
|
Synonyms:
Polidocanol
A687292-100G
A687292-25G
A687292-500G
77669-872EA
10
USD
77669-872
77880-429
77880-430
Polyethylene glycol monododecyl ether Hydroxyl Value 40-60 mg KOH/g
Polyethylene glycol monododecyl ether
Caution: for Research Use Only
Formula:
HO(CH₂CH₂O)nC₁₂H₂₅ |
MDL Number:
MFCD00080891 CAS Number: 9002-92-0 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...