4-Isopropylbenzonitrile ≥98%

Supplier: Ambeed
Warning

A677345-5G A677345-1G
77595-036EA 73.8 USD
77595-036 77749-393
4-Isopropylbenzonitrile ≥98%
4-Isopropylbenzonitrile

Caution: for Research Use Only

Formula: C₁₀H₁₁N
MDL Number: MFCD00052966
CAS Number: 13816-33-6
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR