Penicillin G potassium salt ≥98%

Supplier: Ambeed
Danger

Synonyms: 6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt

A664318-5G A664318-25G
77602-690EA 7.18 USD
77602-690 77669-808
Penicillin G potassium salt ≥98%
Penicillin G potassium salt

Caution: for Research Use Only

Formula: C₁₆H₁₇KN₂O₄S
MW: 372.49 g/mol
MDL Number: MFCD00036193
CAS Number: 113-98-4
UN: 2811
ADR: 6.1 Ⅲ
Merck Index: 12,07225

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR