Ar231453 98+%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: N-(2-Fluoro-4-(methylsulfonyl)phenyl)-6-(4-(3-isopropyl-1,2,4-oxadiazol-5-yl)piperidin-1-yl)-5-nitropyrimidin-4-amine, N-(2-Fluoro-4-(methylsulphonyl)phenyl)-6-(4-(3-isopropyl-1,2,4-oxadiazol-5-yl)piperidin-1-yl)-5-nitropyrimidin-4-amine, AR 231453

A660842-50MG A660842-100MG
77684-552EA 179 USD
77684-552 77886-564
Ar231453 98+%, Ambeed.Inc
Ar231453

Caution: For research use only.

Formula: C₂₁H₂₄FN₇O₅S
MDL Number: MFCD14706714
CAS Number: 733750-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR