3-Methylacetylacetone ≥97%

Supplier: Ambeed
Danger

Synonyms: 3-Methyl-2,4-pentanedione

A653322-1G
77573-094EA 10 USD
77573-094
3-Methylacetylacetone ≥97%
3-Methylacetylacetone

Caution: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00008762
CAS Number: 815-57-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR