N-Ethylisopropylamine ≥98%

Supplier: Ambeed
Danger

A652003-25G A652003-1G A652003-5G
77573-088EA 76 USD
77573-088 77880-369 77880-370
N-Ethylisopropylamine ≥98%
N-Ethylisopropylamine

Caution: for Research Use Only

Formula: C₅H₁₃N
MDL Number: MFCD00015105
CAS Number: 19961-27-4
UN: 2733
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR