1-Pentadecanol ≥98%

Supplier: Ambeed
Warning

A648365-5G A648365-100G
77747-333EA 15 USD
77747-333 77747-332
1-Pentadecanol ≥98%
1-Pentadecanol

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₃CH₂OH
MDL Number: MFCD00004759
CAS Number: 629-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR