Methyl-4-(1-pyrrolidinyl)benzenecarboxylate 98+%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(pyrrolidin-1-yl)benzoate

A597681-1G A597681-5G
77669-580EA 23 USD
77669-580 77880-271
Methyl-4-(1-pyrrolidinyl)benzenecarboxylate 98+%
Methyl-4-(1-pyrrolidinyl)benzenecarboxylate

Caution: for Research Use Only

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD00731259
CAS Number: 129414-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR