Tunicamycin ≥99%

Supplier: Ambeed
Danger

A594348-10MG A594348-5MG
77673-596EA 488 USD
77673-596 77673-598
Tunicamycin ≥99%
Tunicamycin

Caution: for Research Use Only

Formula: C₃₉H₆₄N₄O₁₆
MW: 844.95 g/mol
MDL Number: MFCD00065709
CAS Number: 11089-65-9
UN: 3462
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR